Learn More
Sartorius Vivaflow™ Models Accessories: Flow Restrictors
Flow Restrictors for Vivaflow&trade 50 and 200.
Brand: Sartorius VFA035
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Manage and maintain flow
Specifications
Pink | |
PVC | |
6 Pk. |
Vivaflow 50 and 200 | |
0.6 mm | |
Flow Restrictor |