Learn More
Sartorius Vivaspin™ 15R Centrifugal Concentrators
Features a Hydrosart™ modified regenerated cellulose membrane. Sartorius Vivaspin 15R Centrifugal Concentrators are ideal when extremely high recovery rate and very low adsorption is required, e.g. applications such as concentration and desalting of immunoglobulin fractions. Available in a range of MWCO.
£222.00 - £779.00
Specifications
Material (Membrane) | Hydrosart |
---|---|
Filtration Area | 3.87 cm2 |
Housing Material | Polycarbonate |
Volume (Metric) | 12.5 mL (Fixed Angle Rotor), 15 mL (Swing Bucket Rotor) |
Length (Metric) | 116 mm |
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
10550243
|
Sartorius
VS15RH01 |
10 kDa | 12/Pk. |
£403.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
11765815
|
Sartorius
VS15RH02 |
10 kDa | 48/Pk. |
£779.00
Pack of 48 |
In Stock
Log in to see stock. |
||||
10510823
|
Sartorius
VS15RH21 |
30 kDa | 12/Pk. |
£222.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
10050882
|
Sartorius
VS15RH11 |
5 kDa | 12/Pk. |
£223.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
10117294
|
Sartorius
VS15RH22 |
30 kDa | 48/Pk. |
£779.00
Pack of 48 |
In Stock
Log in to see stock. |
||||
11775815
|
Sartorius
VS15RH12 |
5 kDa | 48/Pk. |
£715.00
Pack of 48 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
11741015
|
Sartorius
VS15RH91 |
2 kDa | 12/Pk. |
£222.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
11735985
|
Sartorius
VS15RH92 |
2 kDa | 48/Pk. |
£779.00
Pack of 48 |
In Stock
Log in to see stock. |
||||
Description
Features a Hydrosart™ modified regenerated cellulose membrane. Sartorius Vivaspin 15R Centrifugal Concentrators are ideal when extremely high recovery rate and very low adsorption is required, e.g. applications such as concentration and desalting of immunoglobulin fractions. Available in a range of MWCO.
The latest member of the Vivaspin product family- With all the unique features of Sartorius Stedim Biotech centrifugal concentrators
- Conical bottom, 30 x 116mm tubes that target a volume of 2 to 15mL
- Patented vertical twin membrane for extremely short concentration time (30-fold in 15 min.)
- Safe sample concentration due to dead-stop volume
- Features the Hydrosart™ regenerated cellulose membrane
- Highest recovery at low absorption (95-98%)
- Very small holdup volume (< 20μl)
- Crystal clear housing with volume graduations
- Designed to fit into rotors that can accommodate Falcon 50mL conical bottom tubes
- Beckman Allegra 25R with TS-5.1-500 swing-out rotor with BUC 5 buckets and 368327 adaptors
- Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
- Multifuge 3 S-R with 75006445 swing out rotor with 75006441 buckets and adaptors for Falcon 50 ml conical bottom tubes
- These devices are not designed to fit into rotors that only accept round bottom 29 mm x 105 mm tubes (e.g. Sorvall SS34 Beckmann JA 20)
- If your rotor accepts only 29 mm x 105 mm round bottom tubes, please use the Vivaspin 15, which can be used in either round bottom or conical centrifuge tubes
Specifications
Hydrosart | |
Polycarbonate | |
116 mm |
3.87 cm2 | |
12.5 mL (Fixed Angle Rotor), 15 mL (Swing Bucket Rotor) |