missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivaspin™ 2 Centrifugal Concentrator with Hydrosart™ Regenerated Cellulose (RC) Membrane
BIOPHARMA

Vivaspin™ 2 Centrifugal Concentrators are disposable ultrafiltration units for concentration of macromolecules. With a broad range of molecular weight cut-offs (MWCO) and membrane materials they are suitable for various substrates, including proteins, viruses, nucleic acids, and nanoparticles.

Brand:  Sartorius™ VS02H01

141.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11778813

  • £212.00 / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Vivaspin concentrators are designed for the concentration or diafiltration of initial sample volumes up to 3 mL and can be used with swinging bucket or fixed angle rotors that accept 15 mL centrifuge tubes. Vivaspin's patented vertical membrane design and thin channel filtration chamber (US 5,647,990) minimize membrane fouling and provide high-speed concentration for even particle-laden solutions. The speed of Vivaspin products combined with low internal surface and membrane area allow superior recovery from very dilute solutions. The Vivaspin 2 also offers flexibility for process optimization: one can directly pipette the concentrate from the dead stop pocket (built into the bottom of the concentrator) or reverse-spin the concentrate into the recovery cap (which can be sealed for storage). Either method results in near-total concentrate recovery.

  • High retentate recovery >90%
  • Vertical membrane design for ultra fast concentration
  • Universal rotor compatibility
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Optional re-spin for concentrate recovery
  • Volume graduation for easy reading/monitoring

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Swinging Bucket Rotor (3 mL), Fixed-angle Rotor (2 mL)
10 kDa
Non-sterile
Centrifugal Concentrator
126 mm
Membrane Filter
Hydrosart
3 mL
25 Pk.
5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius™ Vivaspin™ 2 Centrifugal Concentrator with Hydrosart™ Regenerated Cellulose (RC) Membrane > Vivaspin™ 2 Centrifugal Concentrator, MWCO: 10 kDa, Quantity: 25 Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.