Learn More
Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane
Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.
£197.00 - £1115.00
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | PES |
Filtration Area | 1.3 cm2 |
For Use With (Application) | Concentration, desalting and buffer applications |
Housing Material | Polycarbonate |
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
10384931
|
Sartorius
VS0211 |
5 kDa | 25/Pk. |
£209.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10083930
|
Sartorius
VS0202 |
10 kDa | 100/Pk. |
£1115.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10243281
|
Sartorius
VS0221 |
30 kDa | 25/Pk. |
£209.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10191321
|
Sartorius
VS0201 |
10 kDa | 25/Pk. |
£230.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10527251
|
Sartorius
VS0212 |
5 kDa | 100/Pk. |
£705.00
Pack of 100 |
Estimated Shipment: 31-10-2024
Log in to see stock. |
||||
10551351
|
Sartorius
VS0222 |
30 kDa | 100/Pk. |
£743.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10009450
|
Sartorius
VS0231 |
50 kDa | 25/Pk. |
£209.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10561351
|
Sartorius
VS0232 |
50 kDa | 100/Pk. |
£738.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10596651
|
Sartorius
VS0241 |
100 kDa | 25/Pk. |
£213.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10245471
|
Sartorius
VS0242 |
100 kDa | 100/Pk. |
£1070.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
11748813
|
Sartorius
VS0251 |
300 kDa | 25/Pk. |
£230.00
Pack of 25 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
10609541
|
Sartorius
VS0262 |
1000 kDa | 100/Pk. |
£705.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10708402
|
Sartorius
VS0261 |
1000 kDa | 25/Pk. |
£209.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10153711
|
Sartorius
VS0271 |
25/Pk. |
£209.00
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10357791
|
Sartorius
VS0291 |
3 kDa | 25/Pk. |
£207.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10062741
|
Sartorius
VS0272 |
100/Pk. |
£705.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10302412
|
Sartorius
VS0292 |
3 kDa | 100/Pk. |
£707.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
11730726
|
Sartorius
VS0252 |
300 kDa | 100/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10019780
|
Sartorius
VS02S1 |
5000, 10,000, 30,000, 50,000, 100,000 Da | 25/Pk. (Starter Pack) 5 Ea. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.
Vivaspin concentrators are disposable ultrafiltration devices for concentrating biological samples. Vivaspin 2 is designed for use with samples from 500μl to 2mL in rotors that accept 15mL centrifuge tubes. The unique design provides high-speed concentrations for even particle-laden solutions and near-total concentrate recovery.Twin vertical membranes and optimal design for fast, safe concentration
- Patented twin vertical membrane design for ultra-fast processing speeds
- In a single spin, solution volumes up to 4mL can be concentrated up to 30x in 5–30 minutes
- Thin-channel filtration chamber minimizes membrane fouling and increases filtration speed
- Dead stop pocket built into the bottom eliminates the risk of sample filtration to dryness
- High-flux PES membranes feature very low protein binding and low extractables
- Low internal surface and membrane area for superior recoveries from very dilute solutions
- Two methods for easy concentrate recovery:
- Directly pipet the concentrate from the dead stop pocket in the concentrator
- Reverse-spin the concentrate into the recovery cap, which can be sealed for storage
- Both methods provide near-total concentrate recovery of greater than 90%
- Volume graduation for easy monitoring of concentrate volume and 8μl dead-stop volume
- Universal compatibility with swing bucket or fixed angle rotors accepting 15mL centrifuge tubes
Specifications
Single-use | |
1.3 cm2 | |
Polycarbonate | |
2 mL (Fixed Angle Rotor), 3 mL (Swing Bucket Rotor) | |
17.8 mm |
PES | |
Concentration, desalting and buffer applications | |
Swing bucket rotor: 4,000 x g Fixed angle rotor: 9,000 x g (3 - 50kDa MWCO); 12,000 x g (100kDa+ MWCO) |
|
126 mm |