missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 500 Centrifugal Concentrators
BIOPHARMA

Product Code. 10658961 Shop All Sartorius Products
Click to view available options
MWCO:
0.2 μm
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
5, 10, 30, 50, and 100 kDa
50 kDa
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 10658961

Brand: Sartorius VS0172

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.

  • Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations
  • Concentrate recoveries of > 90% can be reached in less than 30 minutes
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Universal rotor compatibility: can be used in fixed angle rotors accepting 2.2mL centrifuge tubes
  • High-flux PES membranes for ultra-fast concentration
  • Convenient concentrate recovery by pipetting
  • Clear housing with volume graduation for easy reading/monitoring
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
MWCO 0.2 μm
Pore Size 0.2 μm
Sample Volume 500 mL
Sterility Non-sterile
Filtration Area 64.5m m2
Housing Material Polycarbonate
Quantity 100/Pk.
Volume (Metric) 500 μL
Length (Metric) 50 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.