missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 6 Centrifugal Concentrators
BIOPHARMA

Achieve fast sample concentration and high recovery. Sartorius Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.

Brand:  Sartorius VS0692

609.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11770746

  • £920.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Developed to offer increased volume flexibility and performance
  • Compact tubes can process up to an impressive 6mL initial volume
  • Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
  • Universal rotor compatibility:
    • Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
Fast, safe and easy concentration
  • Features premium, high-throughput PES membranes
  • Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Nearly complete concentrate recovery of over 90%
  • Remaining volume is easy to see with the printed graduations and clear housing
  • Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Centrifuge
3 kDa
Non-sterile
Polycarbonate
6 mL
Ultrafiltration
PES
6 mL
2.58 cm2
100/Pk.
122 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.