Learn More
Sartorius Vivaspin™ Turbo 15 PES Centrifugal Concentrators
Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.
Brand: Sartorius VS15T31
171.74 GBP valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Code : 53
Additional Details : Weight : 0.40796kg
Description
Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.
Twin vertical membranes and a sleek internal design for ultra-fast concentration- In a single spin, 15 ml solutions can be concentrated up to 150x in 10–30 minutes
- Large, high-flux PES membranes feature ultra-low protein binding and low extractables
- High, constant filtration pressure allows optimal crossflow to minimize membrane fouling
- Angular, pipette-friendly pocket facilitates easy retentate recovery of greater than 95%
- Eliminates the risk of sample filtration to dryness
- Stable, inert polystyrene housing with high chemical compatibility (pH 1–14)
- UV joining technology for a smooth transition between membrane and housing
- Volume graduation for easy monitoring of concentrate volume and 100μl dead-stop volume
- Handles sample volumes up to 15mL in swing bucket rotors (11mL in fixed angle rotors)
- Designed for rotors that accommodate Falcon 50mL conical tubes:
- Beckman Allegra 25R with TS-5.1-500 swing-out rotor, BUC 5 buckets, adaptors
- Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
- Multifuge 3 S-R with 75006445 rotor, 75006441 buckets, adaptors
Specifications
PES | |
7.2 cm2 | |
Polystyrene | |
12/Pk. | |
11.8 cm |
50 kDa | |
Concentration, desalting or buffer exchange applications | |
4,000 x g | |
11 mL (Fixed Angle Rotor), 15 mL (Swing Bucket Rotor) | |
27 mm |