missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saturated Potassium Chloride Solution, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
250 mL
Description
Glass electrodes need to be kept moist. If an electrode dries out during storage, a regeneration procedure is required to restore the hydrated glass layer and the reference junction in order to make the electrode operable. Filling the storage cap/tube with a potassium chloride solution keeps the electrode hydrated to maximize electrode performance and extend electrode life.
Specifications
Specifications
CAS | 7447-40-7, 7732-18-5 |
Physical Form | Liquid |
Quantity | 250 mL |
Molecular Formula | ClK |
MDL Number | MFCD00011360 |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
Solubility Information | Soluble in water |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
SMILES | [Cl-].[K+] |
IUPAC Name | potassium;chloride |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
EFS424L1 | Certificate of Analysis | 19/02/2025 | 90510 |
EFS422H1 | Certificate of Analysis | 11/12/2024 | 90510 |
EFS422H1 (1) | Certificate of Analysis | 25/10/2022 | 90510 |
147322H1 | Certificate of Analysis | 15/09/2022 | 90510 |
1415501 | Certificate of Compliance | 05/01/2025 | 90510 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Saturated Potassium Chloride Solution, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction