missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories

Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.

Brand:  Thermo Scientific™ SCC5


 View more versions of this product

Product Code. 12642765

  • £1000.16 / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CryoCool Heat Transfer Fluid, 5L
Refrigerated Trap Models RVT100, RVT400, RVT405DDA, RVT5105 and Universal Vacuum Source (UVS) Models UVS400, USV400A, UVS800DDA
5 L
Heat Transfer Fluid
SCC5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to an approved waste disposal plant.

Supplemental information
  • MIXTURE LIST-Contains : octamethytrisiloxane, octamethylcyclotet

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.