missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Medium Capacity Vacuum Concentrators for Combinatorial Chemistry Applications

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

Brand:  Thermo Scientific™ SPD140P2-230

30203.11 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15996171

  • £33903.68 / Each
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

Description

SpeedVac SPD140DDA is a medium capacity vacuum concentrator used with a wide variety of solvents and offer resistance to TFA, DSMO and other aggressive solvents used in combinatorial chemistry applications.   It is available in kit for ease of ordering.

 

SPD140DDA Features:

  • All the product features of SPD130DLX PLUS:
  • Radiant lamps within the chamber
  • Heated lid for processing high boiling points (DMSO,DMF)
  • Independent timers for both heat and run
  • Audible alarm to alert user when operation is finished
  • Built-in vacuum gauge and vacuum display
  • Wider vapor port and tubing for enhanced vapor throughput - reducing drying times
  • Adjustable vacuum level for preventing bumping of difficult solvent mixtures and optimizing drying times
  • -5°C Cold Trap specially designed for trapping high boiling point solvents

Solvent and Solvent Combinations for SPD140P1 KIT

  • Acetic acid
  • Ammonium hydroxide
  • Formic acid
  • Hydrochloric acid
  • TFA
  • Methylene chloride and TFA
  • Acetone and HCL
  • Methylene chloride and MEOH
  • DMSO and methanol

Solvent Combinations for SPD140 P2 KIT

  • DMSO
  • DMF
  • Toluene

SPD140  P1 includes:

  • Acid-Resistant SpeedVac™ Vacuum Concentrator
  • -104°C Ultra-Low Temperature Refrigerated Vapor Trap
  • Oil-Free ETFE/PFA 4-Headed Pump
  • 40 x 1.5-2.0ml Glass Tubes Rotor (RH40-11)
  • Glass Condensation Flask
  • Tubing and Fitting Kit
  • CryoCool™ Heat Transfer Fluid (1L)

SPD140P2 includes:

  • Acid-Resistant SpeedVac™ Vacuum Concentrator
  • -50°C Universal Vacuum Source
  • 24 x 1 dram vials (15 x 45 mm) 4ml - dram vial Rotor (RH24-15)
  • Chemical Trap
  • Tubing and Fitting Kit
  • Disposable Cartridge
  • Glas Condensation Flask
  • Vacuum Pump Lubricating Oil
  • CryoCool™ Heat Transfer Fluid (1L)

SPD140 P1 and P2 kits are recommended for:

  • Solid phase extractions
  • Drying lipid extractions
  • Flash chromatography fractions
  • Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
  • Compound management
  • Evaporating dissolution solventsRotors are available for a wide variety of applications and sample formats. Please check the rotor list.

Typical applications SPD140DDA:

Requires:

  • Drying lipid extraction
  • Flash chromatography fractions
  • Combinatorial Chemistry
  • Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
  • Removing ethyl acetate from samples for CAT assay
  • Drug metabolites in solid phase extracts
  • Analyte elution solvents for evaporation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UL Listed to US and Canadian requirements. CE marked
SpeedVac SPD140 Vacuum Concentrator Kit
230 V 50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers