Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
£29411.45 - £47549.91
Products 3
Product Code Brand Description Electrical Requirements Price Quantity & Availability  
Product Code Brand Description Electrical Requirements Price Quantity & Availability  
15936181
View Documents Promotion Details
Thermo Scientific™
SPD300DDA-230
SpeedVac SPD300DDA Vacuum concentrator 230 V 50 Hz
£29411.45
Each
15946181
View Documents Promotion Details
Thermo Scientific™
SPD300DDAA-230
SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor 230 V 50 Hz
£35598.87
Each
Estimated Shipment: 24-06-2025
to see stock.
Add to basket
15956181
View Documents Promotion Details
Thermo Scientific™
SPD300P1-230
SpeedVac SPD300DDA Vacuum Concentrator Kit 230 V 50 Hz
£47549.91
Each
Description
Description
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
  • For strong acids and bases used in combinatorial chemistry
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits