missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ Universal SpeedVac™ Vacuum System

Safely handle and recover solvents with this single integrated unit, which combines an oil-free vacuum source and refrigerated vapor trap.

Specifications

Shipping Height (English) 18 in.
Shipping Height (Metric) 450 mm
Shipping Length (English) 24 in.
Shipping Length (Metric) 600 mm
Shipping Width (English) 14 in.
View More Specs

Description

Description

Safely handle and recover solvents with the Thermo Scientific™ Savant™ Universal SpeedVac™ Vacuum System, equipped with an easy to use, wide-mouth glass condensation flask. This single integrated unit combines an oil-free vacuum source and refrigerated vapor trap, and can be connected to any of the concentrators to provide a complete SpeedVac system.

Savant UVS400:

  • Low maintenance, oil-free and corrosion resistant system
  • Wide mouth glass condensation flask with self-sealing cover
  • Small footprint
  • Single stage refrigeration system
  • Efficient operation with all SpeedVac concentrators, gel dryers and other vacuum-dependent equipment
  • Optional post trap (model no. ANT100) may be added to neutralize ammonia or acid
  • Chemical trap (model no. DTK120R) may be added to absorb volatile radioactivity and residual vapor
  • Max. vacuum <10 torr (13.3mbar)
  • Oil-free ETFE diaphragm pump
  • 36 liters/minute displacement on 60Hz; 30 liters/minute on 50Hz models

Savant US800DDA:

All the standard features as well as:

  • 3-stage high efficiency oil free pump
  • Max. Vacuum <0.6 torr
  • 36 liters/minute on 60Hz; 20 liters/minute on 50Hz models
  • PTFE tubing and vapor path
  • Post trap ANT100 included
  • Order ANS121/4 ammonia neutralizing solution separately
  • VaporNet™ solvent recovery enhancement

Requires:

Thermo Scientific™ CryoCool™ heat transfer fluid (model no. SCC1) must be ordered for use with all refrigerated cold traps, and the universal vacuum source. Please order separately.

Specifications

Specifications

18 in.
24 in.
14 in.
4 L
450 mm
600 mm
250 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.