missing translation for 'onlineSavingsMsg'
Learn More

SB 202190, 99+%, Thermo Scientific Chemicals

Product Code. 15463629
25 mg
Click to view available options
Quantity:
25 mg
50 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits p38 kinase activity in vivo through competition with ATP

Inhibits p38 kinase activity through competition with ATP

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152121-30-7
Molecular Formula C20H14FN3O, C20H14FN3O
Synonym 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole

Specifications

Quantity 25 mg
Description Yellow
Content And Storage -30°C to -10°C
For Use With (Application) Inhibits p38 kinase activity in vivo through competition with ATP
Formula Weight 331.35g/mol
Melting Point 240°C to 243°C
Solubility Soluble in DMSO at 30mg/ml
Product Type Signal Transduction Reagent
Form Crystalline solid
InChI Key NJNKPVPFGLGHPA-UHFFFAOYSA-N
SMILES FC1=CC=C(C=C1)C1=C(NC(N1)=C1C=CC(=O)C=C1)C1=CC=NC=C1
IUPAC Name 4-[4-(4-fluorophenyl)-5-(pyridin-4-yl)-2,3-dihydro-1H-imidazol-2-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol) 331.35
Assay ≥99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SB 202190, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.