missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals SB 203580, 98%
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Description
Highly specific new inhibitor of p38 kinase and MAP Kinase homologues
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 mg |
| Description | White to Off-white |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | Highly specific new inhibitor of p38 kinase and MAP Kinase homologues |
| Formula Weight | 377.44g/mol |
| Melting Point | 235°C to 247°C |
| Product Type | Signal Transduction Reagent |
| Form | Powder |
| InChI Key | CDMGBJANTYXAIV-UHFFFAOYNA-N |
| SMILES | CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction