missing translation for 'onlineSavingsMsg'
Learn More

SB 366791, Tocris Bioscience™

Product Code. 30001273
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective, competitive TRPV1 antagonist

Potent, selective and competitive vanilloid TRPV1 receptor antagonist (pA2 = 7.71 at hVR1); antagonizes hTRPV1 receptors activated by agonists, noxious heat, but not protons. Displays selectivity over a wide range of receptors and systems including CB1 and CB2 receptors, voltage-gated Ca2+ channels and the hyperpolarization-activated current (Ih).

Chemical Identifiers

CAS 472981-92-3
Molecular Formula C16H14ClNO2
Molecular Weight (g/mol) 287.743
InChI Key RYAMDQKWNKKFHD-JXMROGBWSA-N
Synonym 2e-3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide, n-3-methoxyphenyl-4-chlorocinnamide, n-3-methoxyphenyl-4-chlorocinnamanilide, e-3-4-chlorophenyl-n-3-methoxyphenyl acrylamide, ftalidap 27, 3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide, 2-propenamide, 3-4-chlorophenyl-n-3-methoxyphenyl, sb hplc , powder, e-3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide
PubChem CID 667594
IUPAC Name (E)-3-(4-chlorophenyl)-N-(3-methoxyphenyl)prop-2-enamide
SMILES COC1=CC=CC(=C1)NC(=O)C=CC2=CC=C(C=C2)Cl

Specifications

Quantity 10 mg
Formula Weight 287.75
Percent Purity >99%
Chemical Name or Material SB 366791
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.