missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 415286, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
Potent and selective glycogen synthase kinase-3 (GSK-3) inhibitor (Ki = 31 nM for GSK-3α); competes with ATP. Has minimal activity against 24 other protein kinases (IC50 > 10 μ M). Stimulates glycogen synthesis, gene transcription and is neuroprotective.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 359.73 |
Percent Purity | >99% |
Chemical Name or Material | SB 415286 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'