missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ SC-200 Series Sample Concentrators

Quickly concentrate multiple samples in a block at once.

£1190.00

Specifications

Model SC-200
For Use With (Equipment) Dri-Block™ Inserts
View More Specs

Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17986082
View Documents
Cole-Parmer™
36620-42
For Tubes
£1190.00
1 unit
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
 
17996082
View Documents
Cole-Parmer™
36620-43
For 96-Well Plates
£1190.00
1 unit
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
 
Description

Description

  • Variable needle position suits various Dri-Block™ inserts
  • Small size makes it ideal for use under a fume hood
  • Needles, block, and block heater sold separately
  • Concentrator blows an inert gas over the surface of your samples via stainless steel needles (not included).
  • Blown gas in combination with the heat from the heater produce ideal conditions for fast efficient evaporation.
  • Insert needles into a silicone membrane in any configuration.
  • Fully adjustable gas reservoir provides accurate height control.
  • Maximum vertical traveling distance is 32 cm.
  • Maximum gas usage is 15 L/min.
  • Order PTFE-coated needles for use with corrosive samples.
Specifications

Specifications

SC-200
Dri-Block™ Inserts
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ SC-200 Series Sample Concentrators

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.