missing translation for 'onlineSavingsMsg'
Learn More

SC 26196, Tocris Bioscience™

Product Code. 18074002
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective fatty acid desaturase 2 inhibitor

Selective fatty acid desaturase 2 (FADS2, also known as Δ6 desaturase) inhibitor (IC50 = 0.2 μM in vitro). Displays selectivity over FADS1 and stearoly-CoA 9-desaturase (Δ5 desaturase and SCD-1/Δ9 desaturase, respectively) (IC50 values are >200 μM in vitro). Exhibits anti-inflammatory properties in a mouse edema model. Eliminates CSCs from ovarian cancer cell lines and inhibits sphere formation in vitro.

Chemical Identifiers

CAS 218136-59-5
Molecular Formula C27H29N5
Molecular Weight (g/mol) 423.564
InChI Key QFYKXKMYVYOUNJ-UHFFFAOYSA-N
Synonym α, α-Diphenyl-4-[(3-pyridinylmethylene)amino]-1-piperazinepentanenitrile
PubChem CID 71752132
IUPAC Name 2,2-diphenyl-5-[4-(pyridin-3-ylmethylideneamino)piperazin-1-yl]pentanenitrile
SMILES C1CN(CCN1CCCC(C#N)(C2=CC=CC=C2)C3=CC=CC=C3)N=CC4=CN=CC=C4

Specifications

Quantity 10 mg
Formula Weight 423.55
Percent Purity >98%
Chemical Name or Material SC 26196
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.