missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SC 26196, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective fatty acid desaturase 2 (FADS2, also known as Δ6 desaturase) inhibitor (IC50 = 0.2 μM in vitro). Displays selectivity over FADS1 and stearoly-CoA 9-desaturase (Δ5 desaturase and SCD-1/Δ9 desaturase, respectively) (IC50 values are >200 μM in vitro). Exhibits anti-inflammatory properties in a mouse edema model. Eliminates CSCs from ovarian cancer cell lines and inhibits sphere formation in vitro.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 423.55 |
Percent Purity | >98% |
Chemical Name or Material | SC 26196 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction