missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Scafftag™ Polypropylene Chemtag Inserts
Inserts for the Chemtag™ System. They contain all the pertinent information specific to the risks that hazardous substances pose, such as: substance type, details of hazard and precautions, supplier's name and emergency phone number, etc.
£35.80 - £179.00
Specifications
Product Type | Chemtag Insert |
---|---|
Color | Multicolor |
Material | Polypropylene |
Displayed Text | Chemical Details |
For Use With (Application) | Inspection tagging, Hazard communication Identification |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
12932985
|
Brady™
806687 |
10 ChemTag inserts |
£35.80
Pack of 10 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
12942985
|
Brady™
806683 |
50 ChemTag Inserts |
£179.00
Pack of 50 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
Description
- Contain all the pertinent information specific to the risks that hazardous substances pose, such as:
- Substance type
- Details of hazard and precautions
- Supplier's name and emergency phone number
Specifications
Chemtag Insert | |
Polypropylene | |
Inspection tagging, Hazard communication Identification | |
193 mm | |
-10°C to +90°C | |
5.67 in. |
Multicolor | |
Chemical Details | |
7.6 in. | |
1 Ea. | |
0.5 mm | |
144 mm |