missing translation for 'onlineSavingsMsg'
Learn More

Slice™ Scalpel With Replaceable Blade

Has slim, flat handle and includes finger-friendly™ blade

Brand:  Slice™ 10568

Product Code. 16483639

  • £28.30 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Blade included: straight edge, rounded tip (10518), that's non-magnetic, non-sparking, non-conductive, chemically inert, autoclave-friendly, and rust free
  • Handle lightly weighted, balanced for stability
  • Compatible with all craft blades from Slice™.
  • Finger-friendly™ edge stays sharp 11x longer
  • Reduces injuries, lowers costs
  • Blade safe up to 1600 degrees Celsius
  • Chemically inert blade never rusts
  • No-tool blade change
  • Fewer blade changes = fewer injuries
  • Specifications

    Specifications

    Fixed blade, slim, flat handle (4.1 mm thick)
    5.51 in.
    GFN, Carbon Steel, Polypropylene, Zirconium Oxide
    Safety Cutting
    1 Ea.
    6.34 g
    10 mm
    Scalpel, Precision Knife
    140 mm
    Black/Orange
    1 ceramic straight edge rounded tip blade
    0.23 oz.
    0.39 in.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers