Learn More
Scandium foil, 0.025mm (0.001in) thick, 99% (REO), Thermo Scientific Chemicals
Description
It finds applications in the aerospace industry. Another use of Scandium is in aluminum-scandium alloys for sports equipment, such as bicycle frames, lacrosse sticks, and baseball bats.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon

Specifications
Specifications
Color | Gray |
UN Number | UN3178 |
Quantity | 50 x 50 mm |
Solubility Information | Slowly dissolve in dilute acids. |
Percent Purity | 99.9% |
Odor | Odorless |
Weight | ≈0.05g/25x25mm |
Form | Foil |
Assay | (REO) |
Chemical Name or Material | Scandium foil, 0.025mm (0.001 in.) thick |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
B00X6061 | Certificate of Analysis | 27/01/2023 | 012426.FI,012426.FF |
R12I034 | Certificate of Analysis | 11/08/2022 | 012426.FI,012426.FF |
N21I014 | Certificate of Analysis | 12/04/2022 | 012426.FI,012426.FF |
D09T016 | Certificate of Analysis | 17/11/2021 | 012426.FI,012426.FF |
F02W057 | Certificate of Analysis | 17/11/2021 | 012426.FI,012426.FF |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.