missing translation for 'onlineSavingsMsg'
Learn More

Scandium foil, 0.127mm (0.005in) thick, 99.9% (REO)

Product Code. 11315699
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
This item is not returnable. View return policy

Product Code. 11315699

Brand: Thermo Scientific Alfa Aesar 000294.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 0.24g/25 x 25mm | CAS: 7440-20-2 | Sc | 44.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-20-2
Molecular Formula Sc
Molecular Weight (g/mol) 44.96
MDL Number MFCD00016323
InChI Key SIXSYDAISGFNSX-UHFFFAOYSA-N
Synonym unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
PubChem CID 23952
ChEBI CHEBI:33330
IUPAC Name scandium
SMILES [Sc]

Specifications

Color Silver to Gray
Appearance Silver gray foil
Quantity 50 x 50 mm
Odor Odorless
Assay Percent Range 99.9% (REO)
Form Foil
Chemical Name or Material Scandium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.