missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) 2,4-pentanedionate hydrate

Product Code. p-4408160
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11410760

Brand: Thermo Scientific Alfa Aesar 044448.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 699012-88-9 | C15H21O6Sc | 342.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699012-88-9
Molecular Formula C15H21O6Sc
Molecular Weight (g/mol) 342.28
MDL Number MFCD00064715
InChI Key NYPXMGRKLWIPFL-UHFFFAOYSA-N
Synonym tris 4-hydroxypent-3-en-2-one hydrate scandium
PubChem CID 119030428
IUPAC Name (Z)-4-oxopent-2-en-2-olate;scandium(3+);hydrate
SMILES [Sc+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 174°C to 186°C
Color Colorless
Physical Form Powder
Quantity 1 g
Formula Weight 342.26 (Anhydrous)
Odor Odorless
Chemical Name or Material Scandium(III) 2, 4-pentanedionate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.