missing translation for 'onlineSavingsMsg'
Learn More
Scandium(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11326898

Scandium(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11326898
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11326898

Brand: Thermo Scientific Alfa Aesar 013646.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011501

Specifications

Melting Point >500°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 269.94 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Assay (REO)
Chemical Name or Material Scandium(III) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scandium(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.