missing translation for 'onlineSavingsMsg'
Learn More
Scandium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11333457

Scandium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11333457
5 g
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy

Product Code. 11333457

Brand: Thermo Scientific Alfa Aesar 018677.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scandium(III) chloride is a Prins cyclization catalyst. Used to form an intermetallic nickel-scandium compound film by potentiostatic cathodic reduction from molten salt mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-84-9
MDL Number MFCD00011221

Specifications

Melting Point 939°C
Quantity 5 g
Merck Index 14,8392
Solubility Information Soluble in water; Insoluble in alcohol
Formula Weight 151.32
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 2.39 g/mL
Assay (REO)
Chemical Name or Material Scandium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scandium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.