missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Product Code. 11326268
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11326268

Brand: Thermo Scientific Alfa Aesar 011218.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20662-14-0 | Cl3H12O6Sc | 259.40 g/mol

Scandium(III) chloride hexahydrate, which is freely soluble in water is used as a source for producing metallic scandium by electrolysis. It finds use in halide lamps, optical fibers, electronic ceramics and lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20662-14-0
Molecular Formula Cl3H12O6Sc
Molecular Weight (g/mol) 259.40
MDL Number MFCD00149852
InChI Key FKZFOHABAHJDIK-UHFFFAOYSA-K
IUPAC Name scandium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Sc+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 1 g
Merck Index 14,8392
Solubility Information Soluble in water.
Formula Weight 259.41 (151.32 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Scandium(III) chloride hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.