missing translation for 'onlineSavingsMsg'
Learn More
Scandium(III) iodide, ultra dry, 99.999% (REO), Thermo Scientific Chemicals Product Code.: 11331299

Scandium(III) iodide, ultra dry, 99.999% (REO), Thermo Scientific Chemicals

Product Code. 11331299
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11331299

Brand: Thermo Scientific Alfa Aesar 013687.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scandium iodide is used in high-intensity lighting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14474-33-0
Molecular Formula I3Sc
Molecular Weight (g/mol) 425.669
MDL Number MFCD00054037
InChI Key HUIHCQPFSRNMNM-UHFFFAOYSA-K
Synonym scandium iodide, scandium iii iodide, unii-qvj18sk04t, qvj18sk04t, scandium 3+ triiodide, scandium iii triiodide, acmc-1aq4e
PubChem CID 84464
IUPAC Name scandium(3+);triiodide
SMILES [Sc+3].[I-].[I-].[I-]

Specifications

Melting Point 945°C
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 425.67
Percent Purity 99.999%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Scandium(III) iodide, Ultra dry

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP07K014 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number013687.06,013687.03
Lot NumberU08F006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013687.06,013687.03
Lot NumberT01H011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013687.06,013687.03
Lot NumberT12E052 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number013687.06,013687.03
Lot NumberY30D026 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number013687.06,013687.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scandium(III) iodide, ultra dry, 99.999% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.