missing translation for 'onlineSavingsMsg'
Learn More
Scandium(III) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 11306035

Scandium(III) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 11306035
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11306035

Brand: Thermo Scientific Acros 193960050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-08-1
Molecular Formula O3Sc2
Molecular Weight (g/mol) 137.91
MDL Number MFCD00011223
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
IUPAC Name discandium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]

Specifications

Melting Point >1000.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999%
Linear Formula SC2O3
Merck Index 10,8239
Solubility Information Solubility in water: insoluble
Formula Weight 137.91
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Scandium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467963 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number193960050
Lot NumberA0460211 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number193960010,193960050
Lot NumberA0452183 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number193960050
Lot NumberA0398287 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number193960010,193960050
Lot NumberA0448297 Certificate TypeCertificate of Analysis Date09/12/2022 Catalog Number193960010,193960050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scandium(III) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.