missing translation for 'onlineSavingsMsg'
Learn More
Scandium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11366258

Scandium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11366258
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11366258

Brand: Thermo Scientific Alfa Aesar 011216.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scandium(III) oxide is resistant to heat and thermal shock, hence finds use in high-temperature systems. Further, it is used in electronic ceramics and glass composition. It is used as a starting material for the preparation of other scandium compounds. In addition to this, it has vacuum deposition applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-08-1
Molecular Formula O3Sc2
Molecular Weight (g/mol) 137.91
MDL Number MFCD00011223
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
IUPAC Name discandium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]

Specifications

Melting Point 2485°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water.
Formula Weight 137.91
Odor Odorless
Refractive Index 1.964
Chemical Name or Material Scandium(III) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY14K006 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number011216.06,011216.03,011216.14
Lot Number10252338 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number011216.06,011216.03,011216.14
Lot Number10250963 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number011216.06,011216.03,011216.14
Lot NumberW06J034 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number011216.06,011216.03,011216.14
Lot NumberR20C023 Certificate TypeCertificate of Analysis Date26/05/2022 Catalog Number011216.06,011216.03,011216.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scandium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.