Learn More
Scandium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
Scandium oxide is resistant to heat and thermal shock, hence finds use in high-temperature systems. Further, it is used in electronic ceramics and glass composition. It is used as a starting material for the preparation of other scandium compounds. In addition to this, it has vacuum deposition applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2485°C |
Physical Form | Powder |
Quantity | 1 g |
Assay Percent Range | 99.99% |
Solubility Information | Insoluble in water. |
Formula Weight | 137.91 |
Odor | Odorless |
Refractive Index | 1.964 |
Chemical Name or Material | Scandium(III) oxide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U06K014 | Certificate of Analysis | 30/09/2024 | 011217.06,011217.03,011217.14 |
10251208 | Certificate of Analysis | 30/08/2024 | 011217.06,011217.03,011217.14 |
10251207 | Certificate of Analysis | 30/08/2024 | 011217.06,011217.03,011217.14 |
X11F044 | Certificate of Analysis | 26/09/2022 | 011217.06,011217.03,011217.14 |
F07K24 | Certificate of Analysis | 08/12/2021 | 011217.06,011217.03,011217.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.