missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) oxide, REacton™, 99.998% (metals basis), REM 50ppm max

Product Code. p-4408164
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11381438

Brand: Thermo Scientific Alfa Aesar 010916.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12060-08-1 | O3Sc2 | 137.91 g/mol

A high melting white solid used in high-temperature systems (for its resistance to heat and thermal shock), electronic ceramics, and glass composition. Suitable for vacuum deposition applications. Also used to make scandium fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-08-1
Molecular Formula O3Sc2
Molecular Weight (g/mol) 137.91
MDL Number MFCD00011223
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
IUPAC Name discandium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]

Specifications

Melting Point 2485°C
Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.998% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 137.91
Odor Odorless
Refractive Index 1.964
Chemical Name or Material Scandium(III) oxide, REM 50ppm max.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.