missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

Product Code. 11440311
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11440311

Brand: Thermo Scientific Alfa Aesar 040579.NC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14066-05-8
Molecular Formula Cl3O12Sc
Molecular Weight (g/mol) 343.294
MDL Number MFCD00243289
InChI Key UBVKRMCXDCYZQK-UHFFFAOYSA-K
Synonym scandium perchlorate, scandium 3+ perchlorate, acmc-20ajr3, scandium 3+ ion triperchlorate, trihyperchloric acid scandium salt, scandium iii perchlorate solution, scandium 3+ triperchlorate
PubChem CID 11013318
IUPAC Name scandium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Sc+3]

Specifications

Physical Form Liquid
Quantity 10 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 451.42 (343.30 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Scandium(III) perchlorate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.