Learn More
Scandium ingot, 99.9% (metals basis excluding Ta), Ta nominal 8-12%, Thermo Scientific Chemicals
£141.71 - £520.59
Chemical Identifiers
CAS | 7440-20-2 |
---|---|
Molecular Formula | Sc |
Molecular Weight (g/mol) | 44.96 |
MDL Number | MFCD00016323 |
InChI Key | SIXSYDAISGFNSX-UHFFFAOYSA-N |
Synonym | unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remeltedShow More |
PubChem CID | 23952 |
ChEBI | CHEBI:33330 |
IUPAC Name | scandium |
SMILES | [Sc] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339387
|
Thermo Scientific Alfa Aesar
040229.03 |
1 g |
£141.71
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349387
|
Thermo Scientific Alfa Aesar
040229.06 |
5 g |
£520.59
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7440-20-2 | |
44.96 | |
SIXSYDAISGFNSX-UHFFFAOYSA-N | |
23952 | |
scandium |
Sc | |
MFCD00016323 | |
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remeltedShow More | |
CHEBI:33330 | |
[Sc] |
Specifications
Gray | |
Sc | |
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted | |
[Sc] | |
44.96 | |
CHEBI:33330 | |
Odorless | |
(metals basis excluding Ta), Ta nominal 8 to 12% |
7440-20-2 | |
MFCD00016323 | |
SIXSYDAISGFNSX-UHFFFAOYSA-N | |
scandium | |
23952 | |
99.9% | |
Ingot | |
Scandium ingot |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III
missing translation for 'einecsNumber' : 231-129-2
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.