missing translation for 'onlineSavingsMsg'
Learn More
Scandium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 15441617

Scandium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15441617
2 g, Under Argon
Click to view available options
Quantity:
2 g
Packaging:
Under Argon
This item is not returnable. View return policy

Product Code. 15441617

Brand: Thermo Scientific Alfa Aesar 039996.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-20-2
Appearance Silver gray foil
Density 2.989g/cm3
Assay Percent Range 99.9% (REO)
Molecular Formula Sc
MDL Number MFCD00016323
Quantity 2 g
Synonym unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
InChI Key SIXSYDAISGFNSX-UHFFFAOYSA-N
SMILES [Sc]
IUPAC Name scandium
Molecular Weight (g/mol) 44.96
PubChem CID 23952
ChEBI CHEBI:33330
Odor Odorless
Packaging Under Argon
Chemical Name or Material Scandium, pieces
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scandium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.