Learn More
Merck Scepter™ 3.0 Cell Counter 40 μm Sensors
The Scepter™ 3.0 sensor is engineered with a micro-fabricated, cell-sensing zone enabling discrimination by cell size and volume- down to sub-micron and sub-pL resolution. Coupled with precision liquid handling channels and electronics, the Scepter™ sensor accurately provides cell population statistics.
Brand: Merck PHCC340500
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- For research use only
- Provides fast, convenient method for counting cells and particles
- Compatible with both the 40 and 60 μM sensors
- Employs the Coulter Principle in a miniaturized, handheld, format enabling rapid cell counting, taking less than 30 seconds
- Prepare a dilution of the cell culture of interest and use cell counter to aspirate a sample of this dilution into the sensor
- Displays:
- Cell concentration
- Average cell size
- Average cell volume
- Histogram of size or volume distribution
Spezifikation
Scepter™ 3.0 Cell Counter Sensors | |
50,000 to 1,500,000 particles/mL | |
500 Pk. |
5 to 15 μm | |
4 to 18 μm | |
Cell Counting |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur