Learn More
Merck Scepter™ Test Bead Vial
Scepter™ test beads can be used to assess the Scepter™ cell counter
Brand:  Merck PHCC3BEADS
Product Code. 17191731
  • £251.00 / 5mL
Estimated Shipment: 17-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Scepter™ Test Beads can be used to assess the Scepter™ Cell Counter
  • Compatible with Scepter™ 2.0 and 3.0
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Scepter™ Test Bead Vial
Cell Counting