missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCH 202676 hydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Sulphydryl-reactive compound that inhibits agonist and antagonist binding to G-protein-coupled receptors. Inhibits a variety of GPCRs including adenosine, opioid, muscarinic, adrenergic and dopaminergic receptors (IC50 values are 0.1-1.8 μM).
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 348.26 |
Percent Purity | >99% |
Chemical Name or Material | SCH 202676 hydrobromide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction