missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals SCH772984, 95% Product Code.: 17140656

Thermo Scientific Chemicals SCH772984, 95%

Product Code. 17140656
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 17140656

Brand: Thermo Scientific Chemicals 468271000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 942183-80-4
Assay Percent Range 95%
Quantity 100 mg
Molecular Formula C33H33N9O2
Formula Weight 587.68
InChI Key HDAJDNHIBCDLQF-UHFFFAOYNA-N
SMILES O=C(CN1CCC(C1)C(=O)NC1=CC=C2NN=C(C2=C1)C1=CC=NC=C1)N1CCN(CC1)C1=CC=C(C=C1)C1=NC=CC=N1
IUPAC Name 1-(2-oxo-2-{4-[4-(pyrimidin-2-yl)phenyl]piperazin-1-yl}ethyl)-N-[3-(pyridin-4-yl)-1H-indazol-5-yl]pyrrolidine-3-carboxamide
Molecular Weight (g/mol) 587.69
Chemical Name or Material SCH772984

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals SCH772984, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.