missing translation for 'onlineSavingsMsg'
Learn More
Schiff's Reagent, Pure, for Determination Of Aldehydes, Fisher Chemical™ Product Code.: 10386280

Schiff's Reagent, Pure, for Determination Of Aldehydes, Fisher Chemical™

Product Code. 10386280
1 L
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy

Product Code. 10386280

Brand: Fisher Chemical J/7300/PB15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Schiff's Reagent, Pure, for Determination Of Aldehydes

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 81985

Specifications

Physical Form Liquid
Packaging Plastic Coated Glass bottle
Color Colorless
Quantity 1 L
compliance-icons
Product Identifier
  • Schiff's reagent
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Schiff's Reagent, Pure, for Determination Of Aldehydes, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.