missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Schlenk tubes with NS Stopcock

Nitrogen tubes (Schlenk tubes) with conical ground joint and NS stopcock

£74.75 - £82.80

Specifications

Material Duran Glass
Quantity 1
View More Specs

Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10365981
View Documents
Lenz Laborglasintrumente™
03370213
10 mL
£80.80
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
 
10060191
View Documents
Lenz Laborglasintrumente™
03370228
50 mL
£82.80
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
 
10603442
View Documents
Lenz Laborglasintrumente™
03370137
100 mL
£74.75
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
 
Description

Description

  • With conical ground joint and NS stopcock
  • Material: DURAN™
Specifications

Specifications

Duran Glass
1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente™ Schlenk tubes with NS Stopcock

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.