Learn More
Lenz Laborglasintrumente™ Schlenk Tubes with PTFE Needle-Valve Stopcock
Nitrogen tubes (Schlenk tubes) with conical ground joint and PTFE needle-valve stopcock
£84.85 - £114.00
Specifications
Material Duran Tubing
Pore Size Valve bore 2.5 mm
Quantity 1
View More Specs
Products 8
Product Code Brand Capacity (Metric) Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Type Price Quantity & Availability  
11933421
View Documents
Lenz Laborglasintrumente™
03370813
10 mL Cone NS 14/23
£96.95
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
11953431
View Documents
Lenz Laborglasintrumente™
03370822
25 mL Cone NS 14/23
£96.95
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
11973431
View Documents
Lenz Laborglasintrumente™
03370828
50 mL Cone NS 14/23
£98.95
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
11983431
View Documents
Lenz Laborglasintrumente™
03370837
100 mL Cone NS 14/23
£102.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
11993431
View Documents
Lenz Laborglasintrumente™
03370849
250 mL Cone NS 14/23
£114.00
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
11993411
View Documents
Lenz Laborglasintrumente™
03370622
25 mL Socket NS 14/23
£84.85
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
11903421
View Documents
Lenz Laborglasintrumente™
03370628
50 mL Socket NS 14/23
£85.85
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
11913421
View Documents
Lenz Laborglasintrumente™
03370637
100 mL Socket NS 14/23
£86.85
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
  • With conical ground joint and PTFE needle-valve stopcock
  • Material: DURAN™
Specifications
Specifications
Duran Tubing
Valve bore 2.5 mm
1
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasintrumente™ Schlenk Tubes with PTFE Needle-Valve Stopcock