missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porvair Sciences™ Science Polystyrene Black Microplate
POA - £510.07
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
12449337
|
Porvair Sciences™
223003 |
Non-treated |
£510.07
Pack of 100 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
12394108
|
Porvair Sciences™
223009 |
Tissue Culture |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Square wells maximise liquid volume per well
- White plate prevents cross talk in light emitting assays
- Black plate gives low autofluorescence background
- Volume, total μL/well: 120
- Format: 24 x 16 wells
- Dimensions [w x d x h], mm: 3.75 x 3.75 x 11.4 (well); 85.5 x 127.5 x 14.8 (plate)
Specifications
Standard | |
Black | |
Polystyrene | |
120 μL | |
100 Each |
16 x 24 | |
Microplate | |
384 | |
100 |