missing translation for 'onlineSavingsMsg'
Learn More

Scopolamine hydrobromide, Tocris Bioscience™

Product Code. 30001248
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective muscarinic antagonist

Non-selective muscarinic antagonist. Widely used clinically to treat motion sickness.

Specifications

CAS 114-49-8
Molecular Formula C17H22BrNO4
Quantity 1 g
Synonym scopolamine hydrobromide, unii-1bt935hyhi, 1bt935hyhi, 1alpha,2beta,4beta,5alpha,7beta-1-9-methyl-3-oxa-9-azatricyclo 3.3.1.02,4 non-7-yl hydroxymethyl phenylacetate hydrochloride, 1r,2r,4s,5s,7s-9-methyl-3-oxa-9-azatricyclo 3.3.1.0 2 ,? nonan-7-yl 3-hydroxy-2-phenylpropanoate hydrobromide, 1r,2r,4s,5s,7s-9-methyl-3-oxa-9-azatricyclo 3.3.1.0 2,4 nonan-7-yl 3-hydroxy-2-phenylpropanoate hydrobromide, s-1r,2r,4s,5s,7s-9-methyl-3-oxa-9-azatricyclo 3.3.1.02,4 nonan-7-yl 3-hydroxy-2-phenylpropanoate hydrobromide
InChI Key WTGQALLALWYDJH-BKODDQECSA-N
SMILES CN1C2CC(CC1C3C2O3)OC(=O)C(CO)C4=CC=CC=C4.Br
Molecular Weight (g/mol) 384.27
PubChem CID 51051761
Formula Weight 384.27
Percent Purity >99%
Physical Form Solid
Chemical Name or Material Scopolamine hydrobromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'