missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Scotch™ Contour Tape 471
3M™ Scotch™ Contour Tape 471 is thin, highly flexible, especially stretchy and conformable tape constructed of soft PVC.
Brand: 3M™ 7000028852
Description
- Very easy masking in tight bends
- easy masking even on uneven surfaces
- Good application with waterborne and solventborne paint systems
- sharp paint edges
- Good adhesion even on painted surfaces, glass and rubber
- High flexibility and elasticity
- Good adaptability
- Resistant to many solvents
- Good abrasion resistance
- Multicolor painting or design painting in the car repair area
- Especially for masking plastic parts
- Very fine and clean painting
- Water vapor diffusion 54.3 g / m² x 24 hours

Specifications
Gum Rosin Adhesive | |
Soft PVC | |
12 mm |
Yellow | |
33 m | |
50.8 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Scotch™ Contour Tape 471 > Color: Yellow
Spot an opportunity for improvement?Share a Content Correction