missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Scott™ Control™ Alcohol Foam Hand Sanitizer

Scott™ CONTROL™ alcohol hand sanitizer kills up to 99.99% of microorganisms. Dermatologically tested and fragrance free, foam sanitizer contains aloe vera and skin humectant.

Brand:  Kimberly-Clark™ 6392

75.77 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 17163664

  • £199.00 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Features
    • Contains aloe vera and skin humectant designed to support frequent use
    • Dye and fragrance free formulation is dermatologically tested
    • Quick and easy installation with a simple push and click
    • Empty cassette can be compressed to conserve space and is recyclable once pump is removed
  • Uses:
    • Designed to support compliance control in the workplace
    • Used to minimise risk, support hygiene practices and help contamination control in the workplace
  • Notes:
    • Always read label and product information before use
    • Use biocides safely
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alcohol Foam Hand Sanitizer
Biocidal Products Regulation
Aquarius™ Hand Cleanser Dispensers (6948, 6955, 7124)
6 x 1 L
1 L
Clear
EN 1500, EN 1276, EN13727, EN 14476, EN 1650, EN 13624, BPR Regulation EU 528/2012
Cassette
6 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.