missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Scott™ Control™ Alcohol Foam Hand Sanitizer
Scott™ CONTROL™ alcohol hand sanitizer kills up to 99.99% of microorganisms. Dermatologically tested and fragrance free, foam sanitizer contains aloe vera and skin humectant.
Brand: Kimberly-Clark™ 6393
153.04 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Features
- Contains aloe vera and skin humectant designed to support frequent use
- Dye and fragrance free formulation is dermatologically tested
- Quick and easy installation with a simple push and click
- Empty cassette can be compressed to conserve space and is recyclable once pump is removed
- Uses:
- Designed to support compliance control in the workplace
- Used to minimise risk, support hygiene practices and help contamination control in the workplace
- Notes:
- Always read label and product information before use
- Use biocides safely
Specifications
Alcohol Foam Hand Sanitizer | |
EN 1500, EN 1276, EN13727, EN 14476, EN 1650, EN 13624, BPR Regulation EU 528/2012 | |
Kimberly Clark Professional™ Touchless Electronic Skincare Dispensers in White and Silver | |
4 x 1.2 L | |
1.2 L |
Clear | |
Can be wall mounted or used as freestanding sanitizer dispenser | |
Cassette | |
4.8 L |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction