missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Scott™ Essential™ Examination Couch Paper

Economic and portable, lightweight roll. Scott™ Essential™ Examination Couch Paper ideal for protecting couches in the healthcare industry.

Brand:  Kimberly-Clark Professional™ 7160

50.82 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12600543

  • £159.00 / 1 roll
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Strong, tear-resistant and durable
  • Compatible with dispenser 7056
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Couch Cover
Paper
Single-use
15 in.
15.4 in.
White
38 x 39 cm
Wrapped Roll
38 cm
39 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers