missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Scott™ Performance Small Toilet Tissue Rolls
GREENER_CHOICE

Provides the comforts of home. Scott™ Performance Small Toilet Tissue Rolls dissolves in cold water so it will not clog drains.

Brand:  Kimberly-Clark™ 8538

36.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


Product Code. 12734306

  • £55.50 / 36 rolls
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Fresh white, perforated, 2-ply tissue on small rolls of 320 sheets
  • Helps to control cost and waste
  • Dissolves in cold water to prevent blocked drains
  • ISO 9001 and ISO 14001 accredited
  • Ideal for small office and hotel washrooms especially where economy is important
  • For use with the wall-mounted Aquarius™ Toilet Tissue Dispenser
  • Lower costs per sheet
  • Perforated tissue controls how much is used
  • Manufactured from 100% recycled FSC-certified paper
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Toilet Tissue
12 cm
9.4 cm
ISO 14001, ISO 9001, EU Ecolabel and FSC Certified
12 x 9.4 cm
Ideal for small office and hotel washrooms especially where economy is important
2-ply
4.7 in.
3.7 in.
Paper
White
Roll
6 Packs x 6 Rolls x 320 Sheets (11,520 Sheets)
Small
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.