Learn More
Fisherbrand™ Screen
Accessory for use with vacuum gel dryers
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
Description
- Acts in conjunction with cast-aluminum base
- Serve as porous gel support
- Uniformly distributesthe heat and vacuum
- Prevent gel cracking during drying
- Pore size of stainless steel sheet: 1mm
Specifications
Fisherbrand GD-5040 Gel Dryer System | |
Screen |